Diphenylamine ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-Phenylaniline, N-Phenylbenzeneamine
MDL: MFCD00003014
Beilstein Registry No.: 508755
Notes: ACS melting point specification 52.5-54.0°C. Discolors on exposure to heat.
Soluble in benzene, ether, glacial acetic acid, CS2
CAS No.: 122-39-4
Molecular Formula: C12H11N
Formula Weight: 169.23
Flash Point: 152°(305°F)
Physical Form: Solid
Density: 1.16
Appearance: White to off-white
Melting Point: 52-55°
Boiling Point: 302°
Solubility: Soluble in benzene, ether, glacial acetic acid, CS2
Beilstein Reference: 508755
Merck Reference: 14,3317
UN No.: 3077
MDL No.: MFCD00003014
Formula:
(C₆H₅)₂NH MW: 169.23 g/mol Boiling Pt: 302 Melting Pt: 52…55 °C Density: 1.16 Flash Pt: 152 °C (305 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003014 CAS Number: 122-39-4 EINECS: 204-539-4 UN: 3077 ADR: 9 III Merck Index: 14,3317 |
|
Specification Test Results
Melting point | 52.5-54.0° |
Solubility in alcohol | Pass test |
Residue after ignition | 0.03% |
NO3 | Pass test |
Sensitivity to nitrate | Pass test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...