Aniline 99+% ACS
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aminobenzene, Phenylamine
036238-AP
036238-AE
036238-K2
AA36238-APEA
159.88
USD
AA36238-AP
AA36238-AE
AA36238-K2
Aniline 99+% ACS
Aniline
MDL: MFCD00007629
Beilstein Registry No.: 605631
Fieser: 6,21 17,16
Purity: 99+%
CAS No.: 62-53-3
Molecular Formula: C6H7N
Formula Weight: 93.13
Flash Point: 70°(158°F)
Physical Form: Liquid
Density: 1.021
Refractive Index: 1.5860
Melting Point: -6°
Boiling Point: 183-184°
Beilstein Reference: 605631
Merck Reference: 14,659
UN No.: 1547
MDL No.: MFCD00007629
Formula:
C₆H₅NH₂ MW: 93.13 g/mol Boiling Pt: 183…184 °C Melting Pt: –6 °C Density: 1.021 Flash Pt: 70 °C (158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00007629 CAS Number: 62-53-3 EINECS: 200-539-3 UN: 1547 ADR: 6.1,II Merck Index: 14,659 |
|
Specification Test Results
Color (APHA) | 250 |
Residue after ignition | 0.005% |
C3H5Cl | 0.01% |
Hydrocarbons P.T. | Pass test |
C3H5NO2 | Pass test (limit about 0.001%) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...