Magnesium ≥99.6% (metals basis excluding Ca), Powder -100+200 mesh

Supplier: Thermo Scientific Chemicals
Danger

036194-18 036194-30
AA36194-18EA 106 USD
AA36194-18 AA36194-30
Magnesium ≥99.6% (metals basis excluding Ca), Powder -100+200 mesh
Magnesium

For vacuum deposition


Purity: 99.6% CAS No.: 7439-95-4 Physical Form: Ca typically 0.1-1% Appearance: White Odor: Odorless UN No.: 1418 MDL No.: MFCD00085308


For vacuum deposition

Formula: Mg
MW: 24.3 g/mol
Boiling Pt: 1107 °C (1013 hPa)
Melting Pt: 651 °C
Density: 1,74 g/cm³ (25 °C)
Flash Pt: 500 °C
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1418
ADR: 4.3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR