Cyanogen bromide

Supplier: Thermo Scientific Chemicals
Danger

032731-22 032731-06 032731-14
AA32731-22EA 158 USD
AA32731-22 AA32731-06 AA32731-14
Cyanogen bromide
Cyanogen bromide

MDL: MFCD00011597


CAS No.: 506-68-3 Molecular Formula: BrCN Formula Weight: 105.93 Physical Form: Crystalline Density: 2.015 Odor: Acrid Boiling Point: 61.4° Merck Reference: 14,2693 UN No.: 1889 MDL No.: MFCD00011597

Formula: CBrN
Boiling Pt: 61.4 °C
Melting Pt: 50…52 °C
Density: 2.015
Storage Temperature: Refrigerator
MDL Number: MFCD00011597
CAS Number: 506-68-3
EINECS: 208-051-2
UN: 1889
ADR: 6.1,I
Merck Index: 14,2693

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR