1-Bromopyrene ≥95%

Supplier: Thermo Scientific Chemicals
Warning

018280-03 018280-06
AA18280-03EA 54 USD
AA18280-03 AA18280-06
1-Bromopyrene ≥95%
1-Bromopyrene

MDL: MFCD00015767


Purity: 95% CAS No.: 1714-29-0 Molecular Formula: C16H9Br Formula Weight: 281.13 Physical Form: Crystalline Melting Point: 94-95° MDL No.: MFCD00015767

Formula: C₁₆H₉Br
Melting Pt: 94…95 °C
Storage Temperature: Ambient
MDL Number: MFCD00015767
CAS Number: 1714-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR