Deuterium oxide (99.8% D) (Isotopic)

Supplier: Thermo Scientific Chemicals

014764-18 014764-30
AA14764-18EA 272.96 USD
AA14764-18 AA14764-30
Deuterium oxide (99.8% D) (Isotopic)
Deuterium oxide

MDL: MFCD00044636


Purity: 99.8% CAS No.: 7789-20-0 Molecular Formula: D2O Formula Weight: 20.03 Physical Form: Liquid Density: 1.105 Refractive Index: 1.338 Sensitivity: Moisture Sensitive Odor: Odorless Melting Point: 3.82° Boiling Point: 101.4° Merck Reference: 14,2940 MDL No.: MFCD00044636

Formula: D₂O
Boiling Pt: 101.4 °C
Melting Pt: 3.82 °C
Density: 1.105
Storage Temperature: Ambient
MDL Number: MFCD00044636
CAS Number: 7789-20-0
EINECS: 232-148-9
Merck Index: 14,2940

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR