Sodium azide 99%

Supplier: Thermo Scientific Chemicals
Danger

014314-09 014314-22 014314-36
AA14314-09EA 70.87 USD
AA14314-09 AA14314-22 AA14314-36
Sodium azide 99%
Sodium azide

In airbag inflation, as a preservative in diagnostic medicinals


Purity: 99% CAS No.: 26628-22-8 Molecular Formula: N3Na Formula Weight: 65.01 Physical Form: Powder or Granular Density: 1.846 Appearance: White Odor: Odorless pH: 10 Melting Point: ca 275° dec Solubility: Soluble in water and liquid ammonia. Slightly soluble in alcohol Merck Reference: 14,8581 UN No.: 1687 MDL No.: MFCD00003536


In airbag inflation, as a preservative in diagnostic medicinals

Formula: NaN₃
Melting Pt: ∼ 275 °C (dec.)
Density: 1.846
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1,II
Merck Index: 14,8581

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR