Dysprosium 99,9 REO basis, powder -40 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
012897-14
012897-06
AA12897-14EA
555
USD
AA12897-14
AAA12897-06
Dysprosium 99,9 REO basis, powder -40 mesh
Dysprosium
MDL: MFCD00010982
Purity: 99.9%
CAS No.: 7429-91-6
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00010982
Formula:
Dy MW: 162.5 Density: 8.55 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00010982 CAS Number: 7429-91-6 EINECS: 231-073-9 UN: 3089 ADR: 4.1 II |
Specification Test Results
Form | Packaged under argon |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...