Cesium nitrate ≥99.8% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nitric acid cesium salt
Preparation of cesium compounds
Purity: 99.8%
CAS No.: 7789-18-6
Molecular Formula: CsNO3
Formula Weight: 194.91
Physical Form: Crystalline
Density: 3.68
Refractive Index: 1.55
Sensitivity: Hygroscopic
Appearance: White
Odor: Odorless
Melting Point: 414°
Boiling Point: dec.
Solubility: Soluble in 5 parts cold and 0.5 parts boiling water. Soluble in acetone. Slightly soluble in alcohol
Merck Reference: 14,2015
UN No.: 1451
MDL No.: MFCD00010963
Preparation of cesium compounds
Formula:
CsNO₃ Boiling Pt: Dec. Melting Pt: 414 °C Density: 3.68 Storage Temperature: Ambient |
MDL Number:
MFCD00010963 CAS Number: 7789-18-6 EINECS: 232-146-8 UN: 1451 ADR: 5.1,III Merck Index: 14,2015 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...