Beryllium ≥99.5% (metals basis), irregular lumps 3-38 mm (0.1-1.5 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
012853-18
012853-09
AA12853-18EA
342
USD
AA12853-18
AA12853-09
Beryllium ≥99.5% (metals basis), irregular lumps 3-38 mm (0.1-1.5 in)
Beryllium
MDL: MFCD00134032
Purity: 99.5%
CAS No.: 7440-41-7
Odor: Odorless
MDL No.: MFCD00134032
Formula:
Be Boiling Pt: 2970 °C (1013 hPa) Melting Pt: 1278 °C Density: 1.85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134032 CAS Number: 7440-41-7 EINECS: 231-150-7 Merck Index: 12,01201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...