Triphenylbismuth 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bismuth triphenyle, Triphenylbismuthine

012545-06 012545-14 012545-22
AA12545-06EA 46.53 USD
AA12545-06 AA12545-14 AA12545-22
Triphenylbismuth 99+%
Triphenylbismuth

Volatile organometallic used in the preparation of thin films of superconducting oxides by CVD


Purity: 99+% CAS No.: 603-33-8 Molecular Formula: (C6H5)3Bi Formula Weight: 440.30 Physical Form: Powder Density: 1.585 Appearance: White to off-white Melting Point: 76-81° Boiling Point: 242°/14mm Solubility: Slightly soluble in alcohol. Soluble in ether, acetone and chloroform. Insoluble in water MDL No.: MFCD00014064


Volatile organometallic used in the preparation of thin films of superconducting oxides by CVD

Formula: C₁₈H₁₅Bi
Boiling Pt: 242 °C (14 mmHg)
Melting Pt: 76…81 °C
Density: 1,585
Storage Temperature: Ambient
MDL Number: MFCD00014064
CAS Number: 603-33-8
EINECS: 210-033-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR