Thallium ≥99.99% (metals basis), granules ≤ 6 mm (0.2 in)

Supplier: Thermo Scientific Chemicals
Danger


Packaged in water.

012131-14 012131-22
AA12131-14EA 329 USD
AA12131-14 AA12131-22
Thallium ≥99.99% (metals basis), granules ≤ 6 mm (0.2 in)
Thallium

Purity: 99.99% CAS No.: 7440-28-0 Physical Form: Packaged in water Appearance: Blue-white Odor: Odorless UN No.: 1707 MDL No.: MFCD00134063

Formula: Tl
Boiling Pt: 1457 °C (1013 hPa)
Melting Pt: 301 °C
Density: 11.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134063
CAS Number: 7440-28-0
EINECS: 231-138-1
UN: 1707
ADR: 6.1,II
Merck Index: 12,09391

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR