Potassium nitrate ≥99.994% (metals basis)
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Nitrate of Potash
011008-14
011008-22
AA11008-14EA
86.8
USD
AA11008-14
AA11008-22
Potassium nitrate ≥99.994% (metals basis)
Potassium nitrate
MDL: MFCD00011409
Fieser: 5,561 18,300
Purity: 99.994%
CAS No.: 7757-79-1
Molecular Formula: KNO3
Formula Weight: 101.11
Physical Form: Crystalline
Density: 2.109
Appearance: White
Odor: Odorless
Melting Point: 334°
Boiling Point: 400° dec.
Merck Reference: 14,7648
UN No.: 1486
MDL No.: MFCD00011409
Formula:
KNO₃ MW: 101.1 g/mol Boiling Pt: 400 °C (dec.) Melting Pt: 334 °C Density: 2.109 Storage Temperature: Ambient |
MDL Number:
MFCD00011409 CAS Number: 7757-79-1 EINECS: 231-818-8 UN: 1486 ADR: 5.1,III REACH: 01-2119488224-35 Merck Index: 14,7648 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...