Iron ≥99.995% (metals basis), wire, Puratronic®, Diameter 1.0 mm (0.04 in)

Supplier: Thermo Scientific Chemicals

010935-BW 010935-CB
AA10935-BWEA 181.59 USD
AA10935-BW AA10935-CB
Iron ≥99.995% (metals basis), wire, Puratronic®, Diameter 1.0 mm (0.04 in)
Iron

MDL: MFCD00010999


Purity: 99.995% CAS No.: 7439-89-6 Physical Form: ≈6.2g/m MDL No.: MFCD00010999

Formula: Fe
Boiling Pt: 3000 °C (1013 hPa)
Melting Pt: 1535 °C
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR