Sodium nitrate ≥99.999% (metals basis), Puratronic®

Supplier: Thermo Scientific Chemicals
Danger

010873-22 010873-06 010873-14
AA10873-22EA 851 USD
AA10873-22 AA10873-06 AA10873-14
Sodium nitrate ≥99.999% (metals basis), Puratronic®
Sodium nitrate

MDL: MFCD00011119 Fieser: 18,336


Purity: 99.999% CAS No.: 7631-99-4 Molecular Formula: NaNO3 Formula Weight: 84.99 Physical Form: Powder Density: 2.26 Melting Point: 308° Merck Reference: 14,8647 UN No.: 1498 MDL No.: MFCD00011119

Formula: NaNO₃
MW: 84.99 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 308 °C
Density: 2.26
Storage Temperature: Ambient
MDL Number: MFCD00011119
CAS Number: 7631-99-4
EINECS: 231-554-3
UN: 1498
ADR: 5.1,III
REACH: 01-2119488221-41
Merck Index: 14,8647

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR