Indium ≥99.9995% (metals basis), polycrystalline ingot, Puratronic®
Supplier: Thermo Scientific Chemicals
010616-18
010616-90
AA10616-18EA
387
USD
AA10616-18
AA10616-90
Indium ≥99.9995% (metals basis), polycrystalline ingot, Puratronic®
Indium
MDL: MFCD00134048
Purity: 99.9995%
CAS No.: 7440-74-6
Formula Weight: 114.818
Density: 7.310
Odor: Odorless
Melting Point: 156.6 deg C
Boiling Point: 2072 deg C
MDL No.: MFCD00134048
Formula:
In MW: 114.82 g/mol Boiling Pt: 2000 Melting Pt: 156 Density: 7.310 Storage Temperature: Ambient |
MDL Number:
MFCD00134048 CAS Number: 7440-74-6 EINECS: 231-180-0 Merck Index: 13,04971 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...