Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)

Supplier: Thermo Scientific Chemicals

010614-09 010614-18
AA10614-09EA 177 USD
AA10614-09 AA10614-18
Indium ≥99.9999% (metals basis), shot, Puratronic® ≤ 5 mm (0.2 in)
Indium

MDL: MFCD00134048


Purity: 99.9999+% CAS No.: 7440-74-6 Odor: Odorless MDL No.: MFCD00134048

Formula: In
MW: 114.82 g/mol
Boiling Pt: 2000
Melting Pt: 156
Density: 7.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134048
CAS Number: 7440-74-6
EINECS: 231-180-0
Merck Index: 13,04971

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR