Zinc ≥99% (metals basis), foil 15x15 cm (5.9x5.9 in), Thickness 1.6 mm (0.063 in)

Supplier: Thermo Scientific Chemicals

010438-KG 010438-KJ
AA10438-KGEA 42 USD
AA10438-KG AA10438-KJ
Zinc ≥99% (metals basis), foil 15x15 cm (5.9x5.9 in), Thickness 1.6 mm (0.063 in)
Zinc

MDL: MFCD00011291


Purity: 99% CAS No.: 7440-66-6 Physical Form: ≈255g/15x15cm Odor: Odorless MDL No.: MFCD00011291

Formula: Zn
MW: 65.39 g/mol
Boiling Pt: 908
Melting Pt: 420
Density: 7.14 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011291
CAS Number: 7440-66-6
EINECS: 231-175-3
Merck Index: 12,10255

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR