Magnesium ≥99.8%, powder -325 mesh

Supplier: Thermo Scientific Chemicals
Danger

010233-09 010233-22 010233-57 010233-36
AA10233-09EA 58.3 USD
AA10233-09 AA10233-22 AA10233-57 AA10233-36
Magnesium ≥99.8%, powder -325 mesh
Magnesium

MDL: MFCD00085308


Purity: 99.8% CAS No.: 7439-95-4 Appearance: Silver-grey Odor: Odorless UN No.: 1418 MDL No.: MFCD00085308

Formula: Mg
Storage Temperature: Ambient
MDL Number: MFCD00085308
CAS Number: 7439-95-4
EINECS: 231-104-6
UN: 1418
ADR: 4.3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR