Beryllium ≥99% (metals basis), foil, Thickness 0.1 mm (0.004 in)
Supplier: Thermo Scientific Chemicals
|
Danger
|
010105-FF
010105-FI
AA10105-FFEA
874
USD
AA10105-FF
AA10105-FI
Beryllium ≥99% (metals basis), foil, Thickness 0.1 mm (0.004 in)
Beryllium
MDL: MFCD00134032
Purity: 99.4%
CAS No.: 7440-41-7
Physical Form: ≈0.12g/25x25mm, Vacuum tight
Odor: Odorless
MDL No.: MFCD00134032
Formula:
Be Boiling Pt: 2970 °C (1013 hPa) Melting Pt: 1278 °C Density: 1.85 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134032 CAS Number: 7440-41-7 EINECS: 231-150-7 Merck Index: 12,01201 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...