Lead ≥99% (metals basis), Powder -200 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
000942-22
000942-36
000942-A3
AA00942-22EA
25.3
USD
AA00942-22
AA00942-36
AA00942-A3
Lead ≥99% (metals basis), Powder -200 mesh
Lead
MDL: MFCD00134050
Purity: 99%
CAS No.: 7439-92-1
Odor: Odorless
UN No.: 3077
MDL No.: MFCD00134050
Formula:
Pb MW: 207.2 g/mol Boiling Pt: 1749 °C (1013 hPa) Melting Pt: 327,5 °C Density: 11,3 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00134050 CAS Number: 7439-92-1 EINECS: 231-100-4 UN: 3077 ADR: 9,III |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...