Thallium ≥99.99% (metals basis), rods, Diameter 12.7 mm (0.5 in)

Supplier: Thermo Scientific Chemicals
Danger

000888-0Y 000888-DO
AA00888-0YEA 1689 USD
AA00888-0Y AA00888-DO
Thallium ≥99.99% (metals basis), rods, Diameter 12.7 mm (0.5 in)
Thallium

MDL: MFCD00134063


Purity: 99.99% CAS No.: 7440-28-0 Physical Form: ≈225g/150mm Odor: Odorless MDL No.: MFCD00134063

Formula: Tl
Boiling Pt: 1457 °C (1013 hPa)
Melting Pt: 301 °C
Density: 11.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134063
CAS Number: 7440-28-0
EINECS: 231-138-1
UN: 1707
ADR: 6.1,II
Merck Index: 12,09391

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR