Iron ≥99% (metals basis), powder -200 mesh

Supplier: Thermo Scientific Chemicals
Warning

000737-A1 000737-30
AA00737-A1EA 118 USD
AA00737-A1 AA00737-30
Iron ≥99% (metals basis), powder -200 mesh
Iron

MDL: MFCD00010999


Purity: 99+% CAS No.: 7439-89-6 UN No.: 3089 MDL No.: MFCD00010999

Formula: Fe
MW: 55.85 g/mol
Boiling Pt: 3000
Melting Pt: 1535
Density: 7.86 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010999
CAS Number: 7439-89-6
EINECS: 231-096-4
UN: 3089
ADR: 4.1 III
Merck Index: 13,05109

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR