Copper hard ≥99.999% (metals basis), foil, Puratronic®, hard, Thickness 0.25 mm (0.01 in)

Supplier: Thermo Scientific Chemicals

000097-FY 000097-GD
AA00097-FYEA 184 USD
AA00097-FY AA00097-GD
Copper hard ≥99.999% (metals basis), foil, Puratronic®, hard, Thickness 0.25 mm (0.01 in)
Copper

MDL: MFCD00010965


Purity: 99.999+% CAS No.: 7440-50-8 Physical Form: ≈11.2g/50x100mm, Temper: as rolled Odor: Odorless MDL No.: MFCD00010965

Formula: Cu
MW: 63.55 g/mol
Boiling Pt: 2595
Melting Pt: 1083
Density: 8.94 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00010965
CAS Number: 7440-50-8
EINECS: 231-159-6
Merck Index: 13,02545

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR