Hygromycin B, Ultra Pure Grade
Synonyms:
(3aS,3'R,4S,4'S,5'R,6R,6'R,7S,7aS)-4-(((1R,2S,3R,5S,6R)-3-Amino-2,6-dihydroxy-5-(methylamino)cyclohexyl)oxy)-6'-((S)-1-amino-2-hydroxyethyl)-6-(hydroxymethyl)octahydro-3aH-spiro[[1,3]dioxolo[4,5-c]pyran-2,2'-pyran]-3',4',5',7-tetraol
97064-810EA
109.6
USD
97064-810
Hygromycin B, Ultra Pure Grade
Hygromycin B
An aminoglycoside antibiotic that inhibits protein synthesis in bacteria, fungi and higher eucaryotic cells
Widely used as a selection agent for procaryotic and eucaryotic cells carrying thehygromycin resistance gene. Recommended working concentration 10
Formula:
C₂₀H₃₇N₃O₁₃ MW: 527.5 g/mol |
MDL Number:
MFCD06795479 CAS Number: 31282-04-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...