MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) free acid, Ultra Pure Grade

Supplier: VWR
Warning

J589-100G J589-5KG
97064-266CS 1463.6 USD
97064-266 80760-968 80760-970
MOPSO (3-(4-morpholino)-2-hydroxypropanesulfonic acid) free acid, Ultra Pure Grade
MOPSO (3-(4-Morpholino)-2-hydroxypropanesulphonic acid)

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH. Universally applicable buffers for biochemistry must be water soluble, not interfere with biological processes or biological

Formula: C₇H₁₅NO₅S
Storage Temperature: Ambient
MDL Number: MFCD00009629
CAS Number: 68399-77-9
EINECS: 269-989-6

Order Now

Specification Test Results

Melting Point 276°C - 284°C
Moisture (KF) 1 %
Purity 98 %
Solubility (33.3%, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR