Copper(II) sulfate pentahydrate ACS

Supplier: VWR
Warning

Synonyms: Cupric sulfate pentahydrate, Cupric sulphate pentahydrate, Copper sulphate pentahydrate, Copper sulfate pentahydrate

0330-1KG 0330-500G 0330-2.5KG
97064-980CS 1042.5 USD
97064-980 97064-564 97065-032
Copper(II) sulfate pentahydrate ACS
Copper (II) sulphate pentahydrate
Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Density: 2.284 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
EINECS: 231-847-6
UN: 3077
ADR: 9 III
REACH: 01-2119520566-40
Merck Index: 13,02682

Order Now

Specification Test Results

Calcium 0.005%
Chloride = 0.001%
Insolubles = 0.005%
Iron = 0.003%
Nickel = 0.005%
Nitrogen Compounds (as N) = 0.002%
Potassium = 0.01%
Purity 98% - 102%
Sodium = 0.02%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR