AccessQuick™ RT-PCR System, Promega
Supplier: Promega Corporation
A1701 A1702 A1703
PAA1701EA 220.37 USD
PAA1701 PAA1702 PAA1703
AccessQuick™ RT-PCR System, Promega
Nucleic Acid Reagents RT-PCR and cDNA Reagents
The AccessQuick™ RT-PCR System is an easy and convenient master mix for one-tube RT-PCR.
  • Convenient RT-PCR in One Tube
  • Mix contains Tfl DNA Polymerase, dNTPs, magnesium sulfate and reaction buffer
  • Separate AMV RT provided for important no-RT control reactions
  • Simply combine AccessQuick™ Master Mix with AMV RT, RNA template, primers and water for efficient RT-PCR
The AccessQuick RT-PCR System is an easy and convenient master mix for one-tube RT-PCR. It increases the convenience of performing RT-PCR by combining the following components in a single tube: Tfl DNA Polymerase, dNTPs, magnesium sulfate and reaction buffer. The AMV RT enzyme is provided in a separate tube to allow important no-RT control reactions. The AccessQuick Master Mix is simply added to RNA templates in reaction vials, followed by the AMV RT, primers and water. The AccessQuick RT-PCR Master Mix is intended for routine RT-PCR applications that have been previously optimized and do not require extreme conditions.
Ordering information: Each kit contains AccessQuick™ Master Mix (2X), AMV Reverse Transcriptase and Nuclease-Free Water.