Whatman™ Grade 934-AH™ RTU Glass Microfiber Filters, Whatman products (Cytiva)
Supplier: Cytiva
9907-042 9907-047 9907-055 9907-090 9907-9436 2827-047 3827-042 3827-047 3827-070 3827-090 3827-035 4827-035 4827-042 4827-047 4827-070 4827-090 9927-047 9927-070 9927-090 9907-070
97040-974CS 2279.5 USD
97040-974 97040-976 97040-978 89410-170 75874-050 76097-936 76202-930 76202-932 76202-934 76202-936 76202-940 76202-942 76202-944 76202-946 76202-948 76202-950 76202-952 76202-954 76202-956 76202-958
Whatman™ Grade 934-AH™ RTU Glass Microfiber Filters, Whatman products (Cytiva)
Filters Glass Fiber Filters
These 934-AH™ RTU grade filter papers have been pre-washed, dried (or ignited), cooled, desiccated, and weighed. This eliminates the pre-treating steps that are otherwise required by the US EPA Laboratory Standard Method 2540 parts C, D, and E.
  • Superior fine particle retention at high flow rates
  • Capable of processing very turbid samples
  • Particle retention: 1.5 µm
These filter papers are widely used for dissolved, suspended and volatiles solid analysis. They are also used as a wastewater filter for a wide range of water monitoring applications, including the monitoring of rivers, lakes, and coastal waters, as well as analyzing purified discharge of wastewater treatment plants and monitoring discharge water from industrial plants.
934-AH™ RTU for Suspended and Dissolved Solids: Pre-washed, dried, cooled, and weighed filters with certified mass loss between washing and drying steps of the lesser of less than 0.5 mg or 4% in accordance with US EPA Lab Standard Method 2540 parts C and D. Most filters come weighed to four decimal places, however a version of the 47 mm filter can be measured to five decimal places if needed.
934-AH™ RTU for Volatiles: Pre-washed, pre-weighed, and pre-fired at 550 °C aligned to US EPA Laboratory Standard Method 2540 parts C, D, and E.
934-AH™ RTU Double Weigh: Drying, cooling, and weighing steps repeated twice for customers with the highest regulatory concern.
Packaging: The pre-weighed filter papers are packaged within an aluminium pan with the filter weight clearly printed on a heat resistant label. Each pan has a unique barcode. Each box also has a barcode that can be used to download the weights of all filters from that box.