Eppendorf® 5702/5702 R/5702 RH Benchtop Centrifuge
Supplier: Eppendorf
022628023 022626001 022629905 022626205 022626213 022628001 022628102 022628113 022629883 022629921 022628012 022628124
89203-054EA 10807.66 USD
89203-054 89207-266 47743-472 80076-420 80081-164 97007-364 47745-374 80076-418 80081-166 89207-262 89207-268 89207-270 97007-362 97007-366 89207-264 89207-272 75875-104 75875-106
Eppendorf® 5702/5702 R/5702 RH Benchtop Centrifuge
Centrifuges Benchtop Centrifuges
The range of Eppendorf 5702 general purpose, low-speed centrifuges has been designed specifically to meet the needs of mid-throughput clinical research and cell culture laboratories. These centrifuges are very compact, so that they fit easily on the laboratory bench, as well as extremely quiet.
  • Very compact footprint to fit on every laboratory bench
  • Very quiet operation to benefit your environment
  • Low access height for simple loading and unloading of samples
  • SOFT brake function for slow acceleration and braking. Optimized for cell separation by gradient centrifugation
  • 'At set RPM' function starts the timer when the selected rotational speed is reached for reproducible centrifugation runs
  • Key lock to prevent unintentional adjustments
  • Stainless steel rotor chamber is rust-free and easy to clean
  • Electronic imbalance detection for maximum safety
In addition to the above features centrifuges 5702 R and 5702 RH feature the following:
Fast temprature function for fast precooling.
Continuous cooling maintains a constant temperature after the run has been completed - samples stay cool.
ECO shut-off engages after 8 hours of non-use to reduce energy consumption and extend compressor life.
Two program keys to store routine programs.
Active heating guarantees high temperature accuracy during the entire centrifugation cycle (only 5702 RH).
For in-vitro diagnostic use. The Centrifuge 5702/5702 R/5702 RH is a non-automatic centrifuge for the separation of liquid substance mixtures from the human body and is specifically intended to be used as an accessory together with an in vitro diagnostic device and to enable its use according to its intended purpose.
Make your laboratory environment a better place to work with the 5702 family of low-speed centrifuges that are quiet, user-friendly and so compact that you can fit one comfortably on every lab bench. The low access height allows simple loading and unloading of samples.
The 5702 family offers three different models to suit your laboratory or your application. Centrifuge 5702, which is suitable for standard applications, the refrigerated Centrifuge 5702 R for heat sensitive samples, and the actively heated and refrigerated Centrifuge 5702 RH, which is well suited for working with living specimens, for example in cell culture.
Certifications: UL listed, certificate of quality/conformity (ISO 9001, ISO 13485, ISO 14001).