BioReady Gold Nanospheres, Protein G 40 nm

Supplier: NANOCOMPOSIX, INC.
AUPGR40-1M AUPGR40-10M AUPGR40-200U AUPGR40-5M
77680-122EA 380 USD
77680-122 77680-120 77680-124 77680-126
BioReady Gold Nanospheres, Protein G 40 nm
Beads Microcarriers
40 nm Protein G-conjugated spherical gold nanoparticles combine strong plasmonic optical properties with biofunctionalized surface chemistry. These nanoparticles offer high affinity binding to the Fc region of immunoglobulin G (IgG) from a variety of species. Protein G gold conjugates are used in high sensitivity lateral flow assays, immunoblotting, immunohistochemistry, imaging applications, and more.

  • Solvent: 1X PBS, 0.5% BSA, and 0.05% azide
  • Hydrodynamic diameter (DLS): 50 to 75 nm
  • Zeta potential: ≤–10 mV
  • pH of solution: 6.8 to 8.2
  • Peak wavelength (λmax): 520 to 530 nm

nanoComposix provides high quality nanomaterials that are precisely engineered and highly characterized. All our products are supplied with a batch-specific Certificate of Analysis including characterization data such as TEM, DLS, Zeta, and UV-Vis.

Caution: Intended for Research Use Only.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR