Anti-CD36 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A43697-100UG A43697-50UG
77706-350EA 334.95 USD
77706-350 77706-351
Anti-CD36 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: CD36
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR