Anti-CCKBR Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43690-50UL A43690-100UL
77706-340EA 228.38 USD
77706-340 77706-339
Anti-CCKBR Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, WB:1:500-1:2000

Type:
Antigen: CCKBR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR