Anti-PTPN4 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43684-100UL A43684-50UL
77706-330EA 334.95 USD
77706-330 77706-331
Anti-PTPN4 Rabbit Polyclonal Antibody
Antibodies
Type:
Antigen: PTPN4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR