Anti-CTGF Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A43682-100UG A43682-50UG
77706-327EA 334.95 USD
77706-327 77706-328
Anti-CTGF Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CTGF
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR