Anti-Cxcl1 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A43602-50UG A43602-100UG
77706-187EA 228.38 USD
77706-187 77706-186
Anti-Cxcl1 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: Cxcl1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR