Anti-NPR1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43553-100UL A43553-50UL
77706-098EA 334.95 USD
77706-098 77706-099
Anti-NPR1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:5000, IHC:1:25-1:100

Type:
Antigen: NPR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR