Anti-CIDEC Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A43453-50UL A43453-100UL
77705-918EA 228.38 USD
77705-918 77705-917
Anti-CIDEC Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, IHC:1:25-1:100

Type:
Antigen: CIDEC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR