Anti-SMAD2 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A46089-50UG A46089-100UG
77710-573EA 228.38 USD
77710-573 77710-572
Anti-SMAD2 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SMAD2
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR