Anti-RYR2 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45959-100UG A45959-50UG
77689-206EA 334.95 USD
77689-206 77710-353
Anti-RYR2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200, IF:1:50-1:200

Type:
Antigen: RYR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR