Anti-DDIT4 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45942-100UL A45942-50UL
77710-322EA 334.95 USD
77710-322 77710-323
Anti-DDIT4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: DDIT4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR