Anti-mpt51 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A45884-100UG A45884-50UG
77710-221EA 334.95 USD
77710-221 77710-222
Anti-mpt51 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: mpt51
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycobacterium tuberculosis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR