Anti-pstS1 Rabbit Polyclonal Antibody (HRP)

Supplier: AFG BIOSCIENCE LLC
A45873-100UG A45873-50UG
77710-202EA 334.95 USD
77710-202 77710-203
Anti-pstS1 Rabbit Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: pstS1
Clonality: Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycobacterium tuberculosis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR