Anti-mpt63 Rabbit Polyclonal Antibody (FITC)

Supplier: AFG BIOSCIENCE LLC
A45865-100UG A45865-50UG
77710-189EA 334.95 USD
77710-189 77710-190
Anti-mpt63 Rabbit Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: mpt63
Clonality: Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mycobacterium tuberculosis
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR