Anti-HLA-DQA2 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45826-100UG A45826-50UG
77710-121EA 334.95 USD
77710-121 77710-122
Anti-HLA-DQA2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:10-1:100

Type:
Antigen: HLA-DQA2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR