Anti-HLA-DPA1 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A45823-100UG A45823-50UG
77710-117EA 334.95 USD
77710-117 77710-118
Anti-HLA-DPA1 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: HLA-DPA1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR