Anti-RASSF4 Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45787-50UL A45787-100UL
77710-053EA 228.38 USD
77710-053 77710-052
Anti-RASSF4 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:1000-1:2000, WB:1:200-1:1000

Type:
Antigen: RASSF4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR