Anti-LHB Rabbit Polyclonal Antibody

Supplier: AFG BIOSCIENCE LLC
A45760-100UG A45760-50UG
77710-003EA 334.95 USD
77710-003 77710-004
Anti-LHB Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20-1:200

Type:
Antigen: LHB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR