Anti-IL18R1 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A45545-100UL A45545-50UL
77709-628EA 334.95 USD
77709-628 77709-629
Anti-IL18R1 Rabbit Polyclonal Antibody
Antibodies
ELISA:1:2000-1:5000, IHC:1:50-1:200

Type:
Antigen: IL18R1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR