Anti-AQP3 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG BIOSCIENCE LLC
A45477-50UG A45477-100UG
77709-511EA 228.38 USD
77709-511 77709-510
Anti-AQP3 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: AQP3
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR